Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Map3k9
Entrez Gene ID
338372
Gene Name
mitogen-activated protein kinase kinase kinase 9
Gene Aliases
E130314H24Rik, Mlk1, Prke1
Chromosome Location
Chr.12: 81714950 - 81782549 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001174107.1 NP_001167578.1 7 - 8 1963 72
NM_177395.5 NP_796369.2 7 - 8 1963 72
GenBank mRNA AK053843.1 - 7 - 8 1804 72
AK155677.1 - 7 - 8 1969 72
BC030944.1 - 4 - 5 632 72

View Full Product Details