Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Spag17
Entrez Gene ID
74362
Gene Name
sperm associated antigen 17
Gene Aliases
4931427F14Rik, PF6, Spag17-ps
Chromosome Location
Chr.3: 99885417 - 100143322 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_028892.4 NP_083168.3 20 - 21 2886 112
GenBank mRNA AK029947.1 - 20 - 21 2884 112
AY792594.1 - 20 - 21 2872 112

View Full Product Details