The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001199945.1 | NP_001186874.1 | 8 - 9 | 1229 | 70 |
GenBank mRNA | AK053602.1 | - | 8 - 9 | 1231 | 70 |