The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_007798.3 | NP_031824.1 | 9 - 10 | 1056 | 78 |
GenBank mRNA | AK083393.1 | - | 9 - 10 | 1056 | 78 |
AK147192.1 | - | 9 - 10 | 1003 | 78 | |
AK149884.1 | - | 9 - 10 | 1005 | 78 | |
AK151790.1 | - | 9 - 10 | 1003 | 78 | |
AK153047.1 | - | 9 - 10 | 1002 | 78 | |
AK159992.1 | - | 9 - 10 | 1041 | 78 | |
AK167361.1 | - | 9 - 10 | 1037 | 78 | |
AK170959.1 | - | 9 - 10 | 1029 | 78 | |
BC006656.1 | - | 9 - 10 | 1047 | 78 |