Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Eed
Entrez Gene ID
13626
Gene Name
embryonic ectoderm development
Gene Aliases
ENSMUSG00000039373, l(7)5Rn, l7Rn5, lusk
Chromosome Location
Chr.7: 89954654 - 89981419 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_021876.3 NP_068676.1 4 - 5 880 78
GenBank mRNA AK077664.1 - 4 - 5 465 78
AK087924.1 - 4 - 5 442 78
AK131976.1 - 4 - 5 447 78
AK133890.1 - 4 - 5 441 78
BC012966.1 - 4 - 5 78
U78103.1 - 4 - 5 882 78
U97675.1 - 4 - 5 887 78

View Full Product Details