The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001081123.1 | NP_001074592.1 | 1 - 2 | 216 | 81 |
GenBank mRNA | AK048167.1 | - | 1 - 2 | 104 | 81 |
BC145645.1 | - | 1 - 2 | 182 | 81 |