Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Cenpf
Entrez Gene ID
108000
Gene Name
centromere protein F
Gene Aliases
6530404A22Rik, AI325968, CENF, Lek1, PRO1779, mitosin
Chromosome Location
Chr.1: 189640606 - 189688105 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001081363.2 NP_001074832.2 13 - 14 7970 66
GenBank mRNA AF194970.1 - 2 - 3 6255 66
BC023896.1 - 1 - 2 722 66
DQ642022.1 - 12 - 13 7827 66

View Full Product Details