The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001081363.2 | NP_001074832.2 | 4 - 5 | 605 | 101 |
GenBank mRNA | AK029517.1 | - | 4 - 5 | 607 | 101 |
AK080758.1 | - | 4 - 5 | 592 | 101 | |
AK165236.1 | - | 4 - 5 | 606 | 101 | |
DQ642022.1 | - | 3 - 4 | 462 | 101 |