Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Dbt
Entrez Gene ID
13171
Gene Name
dihydrolipoamide branched chain transacylase E2
Gene Aliases
D3Wsu60e
Chromosome Location
Chr.3: 116513079 - 116549981 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_010022.3 NP_034152.2 8 - 9 1077 69
GenBank mRNA AK075831.1 - 8 - 9 1059 69
AK165959.1 - 8 - 9 1087 69
BC055890.1 - 8 - 9 1033 69
L42996.1 - 8 - 9 1030 69

View Full Product Details