The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001164493.1 | NP_001157965.1 | 13 - 14 | 3180 | 75 |
GenBank mRNA | AK039195.1 | - | 2 - 3 | 281 | 75 |
AK122572.1 | - | 13 - 14 | 2833 | 75 | |
BC065168.1 | - | 13 - 14 | 3180 | 75 | |
BC138283.1 | - | 12 - 13 | 2536 | 75 | |
BC145230.1 | - | 11 - 12 | 2353 | 75 | |
BC145748.1 | - | 12 - 13 | 2536 | 75 |