Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Klhl29
Entrez Gene ID
208439
Gene Name
kelch-like 29
Gene Aliases
A230106N14Rik, Gm68, Kbtbd9, mKIAA1921
Chromosome Location
Chr.12: 5077465 - 5375682 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001164493.1 NP_001157965.1 9 - 10 2479 110
GenBank mRNA AK122572.1 - 9 - 10 2132 110
BC065168.1 - 9 - 10 2479 110
BC138283.1 - 8 - 9 1835 110
BC145748.1 - 8 - 9 1835 110

View Full Product Details