Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Dapp1
Entrez Gene ID
26377
Gene Name
dual adaptor for phosphotyrosine and 3-phosphoinositides 1
Gene Aliases
Bam32
Chromosome Location
Chr.3: 137930998 - 137981576 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_011932.2 NP_036062.1 3 - 4 487 129
GenBank mRNA AF186023.1 - 3 - 4 462 129
AK145745.1 - 3 - 4 453 129
AK146180.1 - 3 - 4 456 129
AK169584.1 - 2 - 3 352 129

View Full Product Details