Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Sec24b
Entrez Gene ID
99683
Gene Name
Sec24 related gene family, member B (S. cerevisiae)
Gene Aliases
AI605202, SEC24
Chromosome Location
Chr.3: 129982764 - 130061573 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_207209.2 NP_997092.1 23 - 24 3886 80
GenBank mRNA AK150076.1 - 22 - 23 2963 80
BC046776.1 - 23 - 24 3886 80
BC057355.1 - 23 - 24 3746 80

View Full Product Details