Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Ift88
Entrez Gene ID
21821
Gene Name
intraflagellar transport 88
Gene Aliases
AW552028, Tg737, Tg737Rpw, TgN737Rpw, Ttc10, flexo, fxo, orpk, polaris
Chromosome Location
Chr.14: 57423657 - 57517936 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_009376.2 NP_033402.2 4 - 5 339 63
GenBank mRNA AK135110.1 - 4 - 5 334 63
AK160633.1 - 4 - 5 348 63
L31959.1 - 4 - 5 284 63

View Full Product Details