Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Exoc5
Entrez Gene ID
105504
Gene Name
exocyst complex component 5
Gene Aliases
AI447711, AI448003, Gm76, PRO1912, SEC10, Sec10l1
Chromosome Location
Chr.14: 49011922 - 49066667 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_207214.3 NP_997097.1 4 - 5 981 133
GenBank mRNA AK134562.1 - 4 - 5 493 133
AK152307.1 - 4 - 5 710 133
AK157473.1 - 4 - 5 676 133
AK164067.1 - 4 - 5 703 133
AK167145.1 - 4 - 5 669 133
AK170148.1 - 4 - 5 674 133
AK171012.1 - 4 - 5 699 133
BC049967.1 - 4 - 5 638 133

View Full Product Details