The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_011372.2 | NP_035502.1 | 4 - 5 | 800 | 134 |
GenBank mRNA | AK034804.1 | - | 4 - 5 | 802 | 134 |
BC058387.1 | - | 4 - 5 | 785 | 134 | |
BC131987.1 | - | 4 - 5 | 751 | 134 | |
BC132317.1 | - | 4 - 5 | 751 | 134 | |
Y11342.1 | - | 4 - 5 | 800 | 134 |