The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_134163.4 | NP_598924.1 | 2 - 3 | 311 | 64 |
GenBank mRNA | AB077700.1 | - | 1 - 2 | 217 | 64 |
AF466292.1 | - | 1 - 2 | 217 | 64 | |
AJ427920.1 | - | 1 - 2 | 217 | 64 | |
AK088971.1 | - | 2 - 3 | 324 | 64 | |
BC057157.1 | - | 2 - 3 | 549 | 64 |