Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Stmn4
Entrez Gene ID
56471
Gene Name
stathmin-like 4
Gene Aliases
Lagl, R75359, RB3
Chromosome Location
Chr.14: 66344303 - 66361680 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_019675.2 NP_062649.1 1 - 2 34 67
GenBank mRNA BC014707.1 - 1 - 2 36 67
BC046752.1 - 1 - 2 34 67

View Full Product Details