The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001277954.1 | NP_001264883.1 | 3 - 4 | 363 | 91 |
NM_001277955.1 | NP_001264884.1 | 3 - 4 | 363 | 91 | |
GenBank mRNA | AK012114.1 | - | - | 305 | 91 |
BC100453.1 | - | - | 208 | 91 |