Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Aen
Entrez Gene ID
68048
Gene Name
apoptosis enhancing nuclease
Gene Aliases
2700083B06Rik, Isg20l1
Chromosome Location
Chr.7: 78895927 - 78908833 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AK076421.1 - - 46 50
CX219175.1 - 1 - 2 162 50

View Full Product Details