Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Picalm
Entrez Gene ID
233489
Gene Name
phosphatidylinositol binding clathrin assembly protein
Gene Aliases
CALM, CLTH, PAP180, fit-1, fit1, mKIAA4114
Chromosome Location
Chr.7: 90130220 - 90209447 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001252520.1 NP_001239449.1 2 - 3 616 141
NM_001252521.1 NP_001239450.1 2 - 3 616 141
NM_001252522.1 NP_001239451.1 2 - 3 616 141
NM_001252523.1 NP_001239452.1 2 - 3 616 141
NM_001252524.1 NP_001239453.1 2 - 3 616 141
NM_146194.4 NP_666306.2 2 - 3 616 141
GenBank mRNA AK144960.1 - 2 - 3 354 141
AK162360.1 - 2 - 3 309 141
AK170373.1 - 2 - 3 636 141
AK220291.1 - 2 - 3 332 141
AY206701.1 - 2 - 3 300 141
BC011470.1 - 2 - 3 326 141
BC021491.1 - 2 - 3 565 141
BC023843.1 - 2 - 3 364 141
BC025566.1 - 2 - 3 639 141
BC057683.1 - 2 - 3 567 141

View Full Product Details