Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Blm
Entrez Gene ID
12144
Gene Name
Bloom syndrome, RecQ helicase-like
Gene Aliases
-
Chromosome Location
Chr.7: 80454993 - 80535119 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001042527.2 NP_001035992.1 12 - 13 2656 126
NM_007550.4 NP_031576.4 12 - 13 2927 126
GenBank mRNA AB008674.1 - 12 - 13 2917 126
AK137022.1 - 12 - 13 2639 126
AK157898.1 - 12 - 13 2657 126
Z98263.1 - 12 - 13 2634 126

View Full Product Details