Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Blm
Entrez Gene ID
12144
Gene Name
Bloom syndrome, RecQ helicase-like
Gene Aliases
-
Chromosome Location
Chr.7: 80454993 - 80535119 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001042527.2 NP_001035992.1 2 - 3 175 114
NM_007550.4 NP_031576.4 2 - 3 446 114
GenBank mRNA AB008674.1 - 2 - 3 436 114
AK137022.1 - 2 - 3 157 114
AK157898.1 - 2 - 3 176 114
AK164605.1 - 2 - 3 144 114
BB614580.1 - 2 - 3 447 114
BB655193.1 - 2 - 3 166 113
Z98263.1 - 2 - 3 153 114

View Full Product Details