Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Atp12a
Entrez Gene ID
192113
Gene Name
ATPase, H+/K+ transporting, nongastric, alpha polypeptide
Gene Aliases
Atp1al1, HKalpha2, cHKA
Chromosome Location
Chr.14: 56365068 - 56388551 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_138652.2 NP_619593.2 20 - 21 3093 66
GenBank mRNA AF100169.1 - 2 - 3 279 66
BC109010.1 - 20 - 21 3091 66
BC109011.1 - 20 - 21 3093 66

View Full Product Details