Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Iars2
Entrez Gene ID
381314
Gene Name
isoleucine-tRNA synthetase 2, mitochondrial
Gene Aliases
2010002H18Rik, C79125
Chromosome Location
Chr.1: 185286662 - 185329419 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_198653.2 NP_941055.1 6 - 7 979 68
GenBank mRNA AK036182.1 - 6 - 7 947 68
AK043590.1 - 6 - 7 981 68
AK043729.1 - 6 - 7 957 68
AK048653.1 - 6 - 7 980 68
AK050358.1 - 6 - 7 962 68
AK051778.1 - 6 - 7 940 68
AK078183.1 - 6 - 7 981 68
AK085714.1 - 6 - 7 976 68
AK088965.1 - 6 - 7 940 68
AK166832.1 - 6 - 7 962 68
BC052403.1 - 6 - 7 956 68

View Full Product Details