Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Rnpc3
Entrez Gene ID
67225
Gene Name
RNA-binding region (RNP1, RRM) containing 3
Gene Aliases
2810441O16Rik, AI447568, C030014B17Rik
Chromosome Location
Chr.3: 113605067 - 113630149 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001038696.1 NP_001033785.1 3 - 4 455 120
NM_001287015.1 NP_001273944.1 3 - 4 455 120
NM_026043.3 NP_080319.2 3 - 4 455 120
GenBank mRNA AK013288.1 - 3 - 4 399 120
AK134241.1 - 3 - 4 420 120
AK161063.1 - 3 - 4 438 120
AK161974.1 - 3 - 4 408 120
AK163729.1 - 3 - 4 408 120
AK173272.1 - 3 - 4 206 120
BC043695.1 - 3 - 4 424 120

View Full Product Details