Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Adamts17
Entrez Gene ID
233332
Gene Name
a disintegrin-like and metallopeptidase (reprolysin type) with thrombospondin type 1 motif, 17
Gene Aliases
AU023434
Chromosome Location
Chr.7: 66839723 - 67153171 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001033877.4 NP_001029049.2 14 - 15 2107 58
GenBank mRNA CN530432.1 - 1 - 2 115 58

View Full Product Details