The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001033877.4 | NP_001029049.2 | 17 - 18 | 2471 | 115 |
GenBank mRNA | BG085304.2 | - | 1 - 2 | 47 | 115 |
CN530432.1 | - | 4 - 5 | 479 | 115 |