Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Larp7
Entrez Gene ID
28036
Gene Name
La ribonucleoprotein domain family, member 7
Gene Aliases
C330027G06Rik, D3Wsu161e
Chromosome Location
Chr.3: 127536714 - 127553349 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_138593.2 NP_613059.2 12 - 13 1801 95
GenBank mRNA AK012726.1 - 1 - 2 103 95
BC029178.1 - 4 - 5 474 95
BC132266.1 - 4 - 5 472 95
BC132268.1 - 4 - 5 472 95
BU701061.1 - 1 - 2 95

View Full Product Details