The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_019398.2 | NP_062271.2 | 1 - 1 | 459 | 117 |
GenBank mRNA | BC117776.2 | - | 1 - 1 | 459 | 117 |
BC119505.1 | - | 1 - 1 | 252 | 117 | |
BC119506.1 | - | 1 - 1 | 255 | 117 |