Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Vps33b
Entrez Gene ID
233405
Gene Name
vacuolar protein sorting 33B (yeast)
Gene Aliases
-
Chromosome Location
Chr.7: 80269635 - 80291579 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_178070.4 NP_835171.2 10 - 11 67
GenBank mRNA AK032076.1 - 10 - 11 67
AK134927.1 - 10 - 11 67
BC011059.1 - 10 - 11 67
BC034170.1 - 10 - 11 817 67

View Full Product Details