The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_178070.4 | NP_835171.2 | 22 - 23 | 2096 | 80 |
GenBank mRNA | AK032076.1 | - | 22 - 23 | 2098 | 80 |
BC011059.1 | - | 22 - 23 | 2088 | 80 | |
BC028783.1 | - | 12 - 13 | 947 | 80 | |
BC034170.1 | - | 22 - 23 | 1808 | 80 |