The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_010375.2 | NP_034505.1 | 4 - 5 | 656 | 89 |
GenBank mRNA | BC099473.1 | - | 4 - 5 | 656 | 89 |
J02872.1 | - | 4 - 5 | 642 | 89 | |
M36900.1 | - | 3 - 4 | 89 | ||
X14092.1 | - | 3 - 4 | 89 |