The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001271468.1 | NP_001258397.1 | 1 - 2 | 234 | 159 |
NM_001271469.1 | NP_001258398.1 | 1 - 2 | 234 | 159 | |
NM_001271492.1 | NP_001258421.1 | 1 - 2 | 234 | 159 | |
NM_010249.2 | NP_034379.1 | 1 - 2 | 234 | 159 | |
NM_207669.2 | NP_997552.1 | 1 - 2 | 234 | 159 | |
NR_073183.1 | 1 - 2 | 234 | 159 | ||
GenBank mRNA | AK040722.1 | - | 1 - 2 | 235 | 159 |
AK052828.1 | - | - | 210 | 159 | |
AK140907.1 | - | 1 - 2 | 145 | 159 | |
M74516.1 | - | 1 - 2 | 129 | 159 | |
M74517.1 | - | 1 - 2 | 128 | 159 |