Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Nedd9
Entrez Gene ID
18003
Gene Name
neural precursor cell expressed, developmentally down-regulated gene 9
Gene Aliases
Cas-L, CasL, HEF1, MEF1, Nedd-9, p105
Chromosome Location
Chr.13: 41309916 - 41487360 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001111324.2 NP_001104794.1 4 - 5 910 79
NM_017464.5 NP_059492.3 3 - 4 691 79
GenBank mRNA AF009366.1 - 4 - 5 846 79
AK030985.1 - 4 - 5 844 79
AK033729.1 - 3 - 4 692 79
AK046357.1 - 3 - 4 654 79
AK054179.1 - 3 - 4 687 79
AK083374.1 - 4 - 5 838 79
BC053713.1 - 4 - 5 843 79

View Full Product Details