The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001267724.1 | NP_001254653.1 | 2 - 3 | 276 | 66 |
NM_178283.4 | NP_840068.1 | 4 - 5 | 615 | 66 | |
GenBank mRNA | AF403041.1 | - | 4 - 5 | 615 | 66 |
AK078289.1 | - | 4 - 5 | 571 | 66 | |
BC018240.1 | - | 4 - 5 | 569 | 66 | |
BG080214.2 | - | 2 - 3 | 210 | 66 |