The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_178283.4 | NP_840068.1 | 5 - 6 | 810 | 84 |
GenBank mRNA | AF403041.1 | - | 5 - 6 | 810 | 84 |
AK078289.1 | - | 5 - 6 | 766 | 84 | |
BC018240.1 | - | 5 - 6 | 764 | 84 |