Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Lce1d
Entrez Gene ID
69611
Gene Name
late cornified envelope 1D
Gene Aliases
2310037L11Rik, AI415320, Sprrl7
Chromosome Location
Chr.3: 92685499 - 92687210 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_027137.2 NP_081413.1 1 - 2 43 65
GenBank mRNA AK009666.1 - 1 - 2 44 65
BC145864.1 - 1 - 2 40 65
BC145866.1 - 1 - 2 40 65

View Full Product Details