The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001081364.3 | NP_001074833.3 | 8 - 9 | 2877 | 82 |
NM_001128084.2 | NP_001121556.2 | 9 - 10 | 2907 | 82 | |
GenBank mRNA | AK122508.1 | - | - | 518 | 82 |
BC076629.1 | - | 8 - 9 | 2877 | 82 | |
BC150741.1 | - | 9 - 10 | 2836 | 82 |