The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001288618.1 | NP_001275547.1 | 8 - 9 | 993 | 64 |
NM_001288619.1 | NP_001275548.1 | 7 - 8 | 900 | 64 | |
NM_001288620.1 | NP_001275549.1 | 6 - 7 | 790 | 64 | |
NM_009953.4 | NP_034083.2 | 8 - 9 | 990 | 64 | |
GenBank mRNA | AK142261.1 | - | 8 - 9 | 940 | 64 |
AY445512.1 | - | 7 - 8 | 900 | 64 | |
AY753668.1 | - | 6 - 7 | 862 | 64 | |
BC137592.1 | - | 8 - 9 | 876 | 64 | |
FJ548565.1 | - | 8 - 9 | 818 | 64 | |
U17858.1 | - | 8 - 9 | 818 | 64 | |
U19939.1 | - | 8 - 9 | 946 | 64 | |
U21729.1 | - | 8 - 9 | 944 | 64 |