Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Slc2a6
Entrez Gene ID
227659
Gene Name
solute carrier family 2 (facilitated glucose transporter), member 6
Gene Aliases
A330096C23, F630103L12Rik, Glut6, Glut9
Chromosome Location
Chr.2: 27021363 - 27028001 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001177627.1 NP_001171098.1 4 - 5 650 73
NM_172659.2 NP_766247.2 4 - 5 650 73
GenBank mRNA AK079650.1 - 4 - 5 594 73
AK089246.1 - 4 - 5 642 73
AK137441.1 - 4 - 5 616 73
AK150104.1 - 4 - 5 603 73
AK163445.1 - 4 - 5 611 73
AK167904.1 - 4 - 5 612 73
AK172411.1 - 4 - 5 594 73
BC058210.1 - 4 - 5 574 73
BC141168.1 - 4 - 5 581 73
BC145480.1 - 4 - 5 581 73

View Full Product Details