Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Slc2a6
Entrez Gene ID
227659
Gene Name
solute carrier family 2 (facilitated glucose transporter), member 6
Gene Aliases
A330096C23, F630103L12Rik, Glut6, Glut9
Chromosome Location
Chr.2: 27021363 - 27028001 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001177627.1 NP_001171098.1 6 - 7 1015 65
NM_172659.2 NP_766247.2 6 - 7 1015 65
GenBank mRNA AK079650.1 - 6 - 7 959 65
AK089246.1 - 6 - 7 1007 65
AK137441.1 - 5 - 6 1230 65
AK150104.1 - 6 - 7 968 65
AK163445.1 - 6 - 7 973 65
AK166312.1 - 6 - 7 973 65
AK172411.1 - 6 - 7 954 65
BC058210.1 - 6 - 7 939 65
BC141168.1 - 6 - 7 946 65
BC145480.1 - 6 - 7 941 65

View Full Product Details