The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001243200.1 | NP_001230129.1 | 3 - 4 | 210 | 66 |
NM_175048.3 | NP_778213.2 | 3 - 4 | 210 | 66 | |
GenBank mRNA | AA543119.1 | - | 3 - 4 | 184 | 66 |
AA795946.1 | - | 3 - 4 | 184 | 66 | |
CB232465.1 | - | 3 - 4 | 241 | 66 |