Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- Prr5l
- Entrez Gene ID
-
72446
- Gene Name
- proline rich 5 like
- Gene Aliases
-
2600010E01Rik, 4833411O04Rik, Protor-2, Protor2
- Chromosome Location
-
Chr.2: 101714280 - 101883074 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
BC172133.1 |
- |
1 - 2 |
247 |
69 |
View Full Product Details