Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Metap1d
Entrez Gene ID
66559
Gene Name
methionyl aminopeptidase type 1D (mitochondrial)
Gene Aliases
2310066F24Rik, 3110033D18Rik, AV117938, MAP 1D, Map1d, MetAP 1D, Metapl1
Chromosome Location
Chr.2: 71453319 - 71525196 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AK139461.1 - - 382 125


Non Gene Transcripts

Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA BB642046.1 - 1 - 2 382 Chr. 2 - 71516558 - 71527919 [-] on Build GRCm38 125


Adjacent Gene Details

Gene Name Gene Symbol Alias Distance Relative Direction Location Chromosome
Upstream histone aminotransferase 1 Hat1 2410071B14Rik, AA536933, Hat-1, KAT1 -76.7kb Anti-sense Chr. - 71389260 - 71441622 on Build GRCm38
Downstream distal-less homeobox 1 Dlx1 Dlx, Dlx-1 122.0 Anti-sense Chr. - 71528041 - 71533981 on Build GRCm38

View Full Product Details