Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Card9
Entrez Gene ID
332579
Gene Name
caspase recruitment domain family, member 9
Gene Aliases
Gm782
Chromosome Location
Chr.2: 26352236 - 26360979 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001037747.1 NP_001032836.1 10 - 11 1429 98
GenBank mRNA BC151023.1 - 10 - 11 1637 98

View Full Product Details