Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Card9
Entrez Gene ID
332579
Gene Name
caspase recruitment domain family, member 9
Gene Aliases
Gm782
Chromosome Location
Chr.2: 26352236 - 26360979 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001037747.1 NP_001032836.1 5 - 6 953 94
GenBank mRNA BC151023.1 - 5 - 6 1161 94

View Full Product Details