Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Slc25a47
Entrez Gene ID
104910
Gene Name
solute carrier family 25, member 47
Gene Aliases
AI132487, AI876593, HDMCP
Chromosome Location
Chr.12: 108849823 - 108856815 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001012310.2 NP_001012310.2 2 - 3 126 69
GenBank mRNA AK149369.1 - 2 - 3 132 69
BC035527.1 - 2 - 3 127 69
BC042739.1 - 1 - 2 254 69
BC055027.1 - 2 - 3 119 69
BC069939.1 - 2 - 3 115 69

View Full Product Details