The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001012310.2 | NP_001012310.2 | 3 - 4 | 194 | 57 |
GenBank mRNA | AK149369.1 | - | 3 - 4 | 200 | 57 |
BC035527.1 | - | 3 - 4 | 195 | 57 | |
BC042739.1 | - | 2 - 3 | 322 | 57 | |
BC055027.1 | - | 3 - 4 | 187 | 57 | |
BC069939.1 | - | 3 - 4 | 183 | 57 | |
BC089616.1 | - | 3 - 4 | 194 | 57 |