Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Atp8b4
Entrez Gene ID
241633
Gene Name
ATPase, class I, type 8B, member 4
Gene Aliases
A530043E15, Im
Chromosome Location
Chr.2: 126320973 - 126500669 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AK086607.1 - 13 - 14 1386 127

View Full Product Details